siRNA Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 4533 | ||
| NM_001322924.1 | NP_001309853.1 | 5 | 4461 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U09197.1 | - | 1 | 161 | Chr. 1 - 167707678 - 167708422 [+] on Build GRCh38 | |

RCSD1
Genomic Map