siRNA Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 4877 | ||
| NM_001322924.1 | NP_001309853.1 | 5 | 4805 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RCSD1| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 4877 | ||
| NM_001322924.1 | NP_001309853.1 | 5 | 4805 | |||