siRNA Details
| Gene Symbol: LOC101929147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125974.1 | 4 | 752 | |||
| GenBank mRNA | BC043601.1 | - | 4 | 723 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | T06091.1 | - | 266 | Chr. - - [-] on Build | ||

LOC101929147
Genomic Map