siRNA Details
Gene Symbol: LOC101929147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125974.1 | 4 | 1801 | |||
GenBank mRNA | BC043601.1 | - | 4 | 1772 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125974.1 | 4 | 1801 | |||
GenBank mRNA | BC043601.1 | - | 4 | 1772 |