siRNA Details
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257273.1 | NP_001244202.1 | 4 | 9303 | ||
XM_011514463.2 | XP_011512765.1 | 3 | 9060 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK023072.1 | - | 1 | 688 | Chr. 6 - 57108788 - 57111032 [+] on Build GRCh38 |
