siRNA Details
| Gene Symbol: ZNF451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514463.2 | XP_011512765.1 | 3 | 9845 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023072.1 | - | 1 | 1473 | Chr. 6 - 57108788 - 57111032 [+] on Build GRCh38 | |

ZNF451
Genomic Map