siRNA Details
| Gene Symbol: ITPKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017001211.1 | XP_016856700.1 | 3 | 8051 | ||
| GenBank mRNA | BC086865.1 | - | 1 | 541 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPKB| Gene Symbol: ITPKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017001211.1 | XP_016856700.1 | 3 | 8051 | ||
| GenBank mRNA | BC086865.1 | - | 1 | 541 | ||