siRNA Details
| Gene Symbol: ASTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648673.1 | - | 1 | 260 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASTN2| Gene Symbol: ASTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648673.1 | - | 1 | 260 | ||