siRNA Details
| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096564.1 | - | 1 | 1434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRK3| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096564.1 | - | 1 | 1434 | ||