siRNA Details
| Gene Symbol: HRAT56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104610.1 | 3, 4 | 2049 | |||
| GenBank mRNA | CR627150.1 | - | 3, 4 | 2049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HRAT56| Gene Symbol: HRAT56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104610.1 | 3, 4 | 2049 | |||
| GenBank mRNA | CR627150.1 | - | 3, 4 | 2049 | ||