siRNA Details
| Gene Symbol: LINC01560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126059.1 | 1 | 940 | |||
| GenBank mRNA | BC025179.1 | - | 1 | 85 | ||
| HY147632.1 | - | 1 | 144 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI260296.1 | - | 1 | 86 | Chr. X - 47483571 - 47484361 [+] on Build GRCh38 | |

LINC01560
Genomic Map