siRNA Details
| Gene Symbol: LINC01560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126059.1 | 1 | 1155 | |||
| GenBank mRNA | BC025179.1 | - | 1 | 300 | ||
| HY147632.1 | - | 1 | 359 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA278227.1 | - | 1 | 16 | Chr. X - 47483855 - 47484331 [+] on Build GRCh38 | |

LINC01560
Genomic Map