siRNA Details
| Gene Symbol: FGFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094303.1 | - | 1 | 331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR1| Gene Symbol: FGFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094303.1 | - | 1 | 331 | ||