siRNA Details
| Gene Symbol: FGFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094303.1 | - | 1 | 1705 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA646574.1 | - | 1 | 192 | Chr. 8 - 38445342 - 38446131 [-] on Build GRCh38 | |

FGFR1
Genomic Map