siRNA Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142339.2 | NP_001135811.1 | 13 | 5698 | ||
| NM_001261443.1 | NP_001248372.1 | 13 | 5500 | |||
| NM_001261444.1 | NP_001248373.1 | 8 | 5218 | |||
| NM_182978.3 | NP_892023.1 | 12 | 6141 | |||

GNAL
Genomic Map