siRNA Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142339.2 | NP_001135811.1 | 13 | 4349 | ||
| NM_001261443.1 | NP_001248372.1 | 13 | 4151 | |||
| NM_001261444.1 | NP_001248373.1 | 8 | 3869 | |||
| NM_182978.3 | NP_892023.1 | 12 | 4792 | |||
| Gene Symbol: MPPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL536593.3 | - | 1 | 681 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR593590.1 | - | 1 | 681 | Chr. 18 - 11883321 - 11885649 [+] on Build GRCh38 | |

GNAL
Genomic Map