siRNA Details
| Gene Symbol: PPARGC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513770.2 | XP_011512072.1 | 1 | 322 | ||
| GenBank mRNA | HQ695733.1 | - | 1 | 322 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK095100.1 | - | 1 | 1003 | Chr. 4 - 23879202 - 23881973 [-] on Build GRCh38 | |

PPARGC1A
Genomic Map