siRNA Details
| Gene Symbol: TIMM17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029446.1 | - | 1 | 214 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG542570.1 | - | 1 | 215 | Chr. X - 48897158 - 48897781 [-] on Build GRCh38 | |

TIMM17B
Genomic Map