siRNA Details
| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005248218.4 | XP_005248275.1 | 5 | 3960 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK001995.1 | - | 1 | 1978 | Chr. 4 - 13456451 - 13458546 [-] on Build GRCh38 | |

RAB28
Genomic Map