siRNA Details
| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092074.1 | - | 1 | 1693 | ||
| AK094997.1 | - | 1 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM212B| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092074.1 | - | 1 | 1693 | ||
| AK094997.1 | - | 1 | 381 | |||