siRNA Details
| Gene Symbol: LOC101928933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC039325.1 | - | 1 | 690 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928933| Gene Symbol: LOC101928933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC039325.1 | - | 1 | 690 | ||