siRNA Details
| Gene Symbol: LOC101927310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110756.1 | 1 | 244 | |||
| GenBank mRNA | BC043294.2 | - | 1 | 244 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000559363.2 | - | 1 | 248 | Chr. 15 - 97553284 - 97560702 [-] on Build GRCh38 | |

LOC101927310
Genomic Map