siRNA Details
| Gene Symbol: LOC101927310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110756.1 | 1 | 497 | |||
| GenBank mRNA | BC043294.2 | - | 1 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927310| Gene Symbol: LOC101927310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110756.1 | 1 | 497 | |||
| GenBank mRNA | BC043294.2 | - | 1 | 497 | ||