siRNA Details
| Gene Symbol: STRBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014890.1 | XP_016870379.1 | 17 | 4428 | ||
| XM_017014891.1 | XP_016870380.1 | 17 | 4401 | |||
| XM_017014892.1 | XP_016870381.1 | 17 | 4660 | |||
| XM_017014893.1 | XP_016870382.1 | 17 | 4370 | |||

STRBP
Genomic Map