siRNA Details
| Gene Symbol: STRBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014890.1 | XP_016870379.1 | 17 | 4637 | ||
| XM_017014891.1 | XP_016870380.1 | 17 | 4610 | |||
| XM_017014892.1 | XP_016870381.1 | 17 | 4869 | |||
| XM_017014893.1 | XP_016870382.1 | 17 | 4579 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024960.1 | - | 1 | 1775 | Chr. 9 - 123125407 - 123127550 [-] on Build GRCh38 | |

STRBP
Genomic Map