siRNA Details
| Gene Symbol: LOC389273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134289.1 | 3 | 842 | |||
| NR_134290.1 | 3 | 651 | ||||
| GenBank mRNA | AK130861.1 | - | 3 | 651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC389273| Gene Symbol: LOC389273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134289.1 | 3 | 842 | |||
| NR_134290.1 | 3 | 651 | ||||
| GenBank mRNA | AK130861.1 | - | 3 | 651 | ||