siRNA Details
| Gene Symbol: SH3TC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC068094.1 | - | 3 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3TC1| Gene Symbol: SH3TC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC068094.1 | - | 3 | 293 | ||