siRNA Details
| Gene Symbol: ZNF281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 4704 | ||
| NM_001281294.1 | NP_001268223.1 | 3 | 4596 | |||
| NM_012482.4 | NP_036614.1 | 2 | 4700 | |||
| XM_017000887.1 | XP_016856376.1 | 5 | 6085 | |||
| XM_017000888.1 | XP_016856377.1 | 5 | 5043 | |||
| GenBank mRNA | BC016000.1 | - | 1 | 972 | ||

ZNF281
Genomic Map