siRNA Details
Gene Symbol: LOC101928063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110727.1 | 2 | 1138 | |||
GenBank mRNA | AK090553.1 | - | 2 | 746 | ||
AK094825.1 | - | 2 | 1101 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110727.1 | 2 | 1138 | |||
GenBank mRNA | AK090553.1 | - | 2 | 746 | ||
AK094825.1 | - | 2 | 1101 |