siRNA Details
| Gene Symbol: PRO2964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF119910.1 | - | 1 | 340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRO2964| Gene Symbol: PRO2964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF119910.1 | - | 1 | 340 | ||