siRNA Details
| Gene Symbol: MLYCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094095.1 | - | 1 | 116 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124050.1 | - | 1 | 1494 | Chr. 16 - 83911683 - 83913803 [+] on Build GRCh38 | |

MLYCD
Genomic Map