siRNA Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 5353 | ||
| GenBank mRNA | AK024927.1 | - | 1 | 839 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG743165.1 | - | 1 | 257 | Chr. 6 - 54943952 - 54944665 [+] on Build GRCh38 | |

FAM83B
Genomic Map