siRNA Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 5553 | ||
| GenBank mRNA | AK024927.1 | - | 1 | 1039 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM83B| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 5553 | ||
| GenBank mRNA | AK024927.1 | - | 1 | 1039 | ||