siRNA Details
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 5512 | ||
GenBank mRNA | AK024927.1 | - | 1 | 998 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010872.2 | NP_001010872.1 | 5 | 5512 | ||
GenBank mRNA | AK024927.1 | - | 1 | 998 |