siRNA Details
Gene Symbol: PARTICL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038942.1 | 1 | 663 | |||
GenBank mRNA | BC009563.1 | - | 1 | 663 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PARTICL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038942.1 | 1 | 663 | |||
GenBank mRNA | BC009563.1 | - | 1 | 663 |