siRNA Details
| Gene Symbol: PARTICL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038942.1 | 1 | 1249 | |||
| GenBank mRNA | BC009563.1 | - | 1 | 1249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARTICL| Gene Symbol: PARTICL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038942.1 | 1 | 1249 | |||
| GenBank mRNA | BC009563.1 | - | 1 | 1249 | ||