siRNA Details
| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303456.1 | NP_001290385.1 | 15 | 3219 | ||
| NM_006468.7 | NP_006459.3 | 15 | 3417 | |||
| GenBank mRNA | BQ932484.1 | - | 1 | 896 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR3C| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303456.1 | NP_001290385.1 | 15 | 3219 | ||
| NM_006468.7 | NP_006459.3 | 15 | 3417 | |||
| GenBank mRNA | BQ932484.1 | - | 1 | 896 | ||