siRNA Details
| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303456.1 | NP_001290385.1 | 15 | 3441 | ||
| NM_006468.7 | NP_006459.3 | 15 | 3639 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR606271.1 | - | 1 | 920 | Chr. 1 - 145843217 - 145844402 [+] on Build GRCh38 | |

POLR3C
Genomic Map