siRNA Details
| Gene Symbol: ADRA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648936.1 | - | 1 | 3535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRA1B| Gene Symbol: ADRA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648936.1 | - | 1 | 3535 | ||