siRNA Details
| Gene Symbol: LINC01278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023798.1 | - | 1 | 1196 | ||
| AK130294.1 | - | 1 | 2322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01278| Gene Symbol: LINC01278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023798.1 | - | 1 | 1196 | ||
| AK130294.1 | - | 1 | 2322 | |||