siRNA Details
| Gene Symbol: MGC10814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC004943.1 | - | 1 | 430 | 3613095 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC10814| Gene Symbol: MGC10814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC004943.1 | - | 1 | 430 | 3613095 | |