siRNA Details
Gene Symbol: MGC10814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004943.1 | - | 11 | 3613095 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX283468.1 | - | 12 | Chr. 19 - 18881599 - 18882126 [-] on Build GRCh38 |
