siRNA Details
| Gene Symbol: NDUFS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056061.1 | - | 1 | 461 | ||
| AK091024.1 | - | 6 | 1325 | |||
| AK091046.1 | - | 1 | 2018 | |||
| AK091623.1 | - | 7 | 1096 | |||
| AK092169.1 | - | 6 | 1364 | |||
| AK128713.1 | - | 5 | 1631 | |||
| AK297611.1 | - | 6 | 1168 | |||

NDUFS7
Genomic Map