siRNA Details
| Gene Symbol: LINC01262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121679.1 | 3 | 703 | |||
| GenBank mRNA | BC092460.1 | - | 3 | 703 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000513681.1 | - | 3 | 704 | Chr. 4 - 189659605 - 189661486 [+] on Build GRCh38 | |

LINC01262
Genomic Map