siRNA Details
| Gene Symbol: LINC01262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121679.1 | 1, 2 | 134 | |||
| GenBank mRNA | BC092460.1 | - | 1, 2 | 134 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD641470.1 | - | 137 | Chr. - - [-] on Build | ||

LINC01262
Genomic Map