siRNA Details
| Gene Symbol: LINC01262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC092460.1 | - | 3 | 583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01262| Gene Symbol: LINC01262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC092460.1 | - | 3 | 583 | ||