siRNA Details
| Gene Symbol: RASGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL359931.1 | - | 1 | 1299 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA119282.1 | - | 1 | 20 | Chr. 15 - 78979837 - 78980387 [-] on Build GRCh38 | |

RASGRF1
Genomic Map