siRNA Details
| Gene Symbol: SYTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056722.1 | - | 1 | 1038 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL700484.1 | - | 1 | 144 | Chr. 11 - 85705220 - 85705872 [-] on Build GRCh38 | |

SYTL2
Genomic Map