siRNA Details
| Gene Symbol: NCBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114118.2 | NP_001107590.1 | 13 | 6489 | ||
| XM_005256717.2 | XP_005256774.1 | 14 | 6027 | |||
| XM_006721556.2 | XP_006721619.1 | 13 | 5916 | |||
| XM_011523954.2 | XP_011522256.1 | 14 | 5944 | |||
| XM_011523955.2 | XP_011522257.1 | 13 | 5798 | |||
| XM_017024827.1 | XP_016880316.1 | 8 | 5501 | |||
| XM_017024828.1 | XP_016880317.1 | 9 | 5319 | |||
| GenBank mRNA | AK056724.1 | - | 1 | 409 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA284923.1 | - | 1 | 39 | Chr. 17 - 3808033 - 3808484 [-] on Build GRCh38 | |

NCBP3
Genomic Map