siRNA Details
| Gene Symbol: NCBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114118.2 | NP_001107590.1 | 13 | 7314 | ||
| XM_005256717.2 | XP_005256774.1 | 14 | 6852 | |||
| GenBank mRNA | AK056724.1 | - | 1 | 1234 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG675548.1 | - | 1 | 576 | Chr. 17 - 3807416 - 3808194 [-] on Build GRCh38 | |

NCBP3
Genomic Map